missing translation for 'onlineSavingsMsg'
Learn More

Carbuterol Hemisulfate Salt, TRC

Product Code. 30357844
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357844 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30357844 Supplier Toronto Research Chemicals Supplier No. TRCC18335010MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Carbuterol Hemisulfate Salt
Purity Grade Notes HPLC
Molecular Formula 2 C13 H21 N3 O3 . H2 O4 S
Quantity 10 mg
Synonym Carbuterol Hemisulfate, Urea, N-[5-[2-[(1,1-dimethylethyl)amino]-1-hydroxyethyl]-2-hydroxyphenyl]-, sulfate (2:1), N-[5-[2-[(1,1-Dimethylethyl)amino]-1-hydroxyethyl]-2-hydroxyphenyl]urea Hemisulfate, Bronsecur, Carbuterol Hemisulfate Pirem, SKF 40383 Hemisulfate, SKF 40383A Hemisulfate
InChI Formula InChI=1S/2C13H21N3O3.H2O4S/c2*1-13(2,3)15-7-11(18)8-4-5-10(17)9(6-8)16-12(14)19;1-5(2,3)4/h2*4-6,11,15,17-18H,7H2,1-3H3,(H3,14,16,19);(H2,1,2,3,4)
SMILES CC(C)(C)NCC(O)c1ccc(O)c(NC(=O)N)c1.CC(C)(C)NCC(O)c2ccc(O)c(NC(=O)N)c2.OS(=O)(=O)O
IUPAC Name [5-[2-(tert-butylamino)-1-hydroxyethyl]-2-hydroxyphenyl]urea;sulfuric acid
Molecular Weight (g/mol) 632.7268
Formula Weight 632.284
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.