missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrum Labs™ Carboy Process Reservoir with Drain
Click to view available options
Volume (Metric):
20 L
50 L
Description
- Tubes are cut and positioned to ensure that all of solution is directed into the module, and to prevent foaming
- Reservoirs have been intentionally crafted for seamless integration into SpectrumLabs™ TFF pump systems
Specifications
Specifications
| Material | Polypropylene |
| For Use With (Application) | KMPi systems |
| Includes | 3/4 in. TC x 43 bent dip tubes3/8 in. HB Vent (Green)3/4 in. TC Inlet (Orange)3/4 in. TC Outlet (Blue)3/4 in. TC Buffer (Yellow)3/4 in. TC Drain3 in. TC Bottle Opening |
| Type | Carboy TFF Reservoir |
| Volume (Metric) | 20 L |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction