Learn More
Spectrum Labs™ Carboy Process Reservoir with Drain
Product Code. 15826511
Click to view available options
Volume (Metric):
20 L
50 L
Product Code. 15826511
Brand: Spectrum Labs™ ACBT20TC
This item requires a price update prior to being ordered.
Request a Price for this item here.
Spectrum Labs™ Carboy Process Reservoir with Drain are designed with a combination of configured ports and tubes to support tangential flow filtration (TFF) applications.
  • Tubes are cut and positioned to ensure that all of solution is directed into the module, and to prevent foaming
  • Reservoirs have been intentionally crafted for seamless integration into SpectrumLabs™ TFF pump systems
Specifications
Material Polypropylene
For Use With (Application) KMPi systems
Includes 3/4 in. TC x 43 bent dip tubes3/8 in. HB Vent (Green)3/4 in. TC Inlet (Orange)3/4 in. TC Outlet (Blue)3/4 in. TC Buffer (Yellow)3/4 in. TC Drain3 in. TC Bottle Opening
Type Carboy TFF Reservoir
Volume (Metric) 20 L