missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carboxypeptidase M Antibody (OTI2D11), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70446PCP
This item is not returnable.
View return policy
Description
Carboxypeptidase M Monoclonal antibody specifically detects Carboxypeptidase M in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| Carboxypeptidase M | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 1368 | |
| Store at 4C in the dark. | |
| IgG2b |
| Western Blot, Immunohistochemistry | |
| OTI2D11 | |
| Western Blot, Immunohistochemistry | |
| carboxypeptidase M, EC 3.4.17, EC 3.4.17.12 | |
| Human recombinant protein fragment corresponding to amino acids 124-423 of human CPM(NP_001865) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction