missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carboxypeptidase M Antibody (105), Alexa Fluor™ 488, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89359AF488
This item is not returnable.
View return policy
Description
Carboxypeptidase M Monoclonal specifically detects Carboxypeptidase M in Human samples. It is validated for Western Blot, ELISA.
Specifications
| Carboxypeptidase M | |
| Monoclonal | |
| Alexa Fluor 488 | |
| carboxypeptidase M, EC 3.4.17, EC 3.4.17.12 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Carboxypeptidase M/ CPM (NP_938079.1; Met 1-His 422). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 105 | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 1368 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction