missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Carboxypeptidase B1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA528204

Product Code. 13412317

  • £450.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Recommended positive controls: AsPC-1, mouse pancreas, rat pancreas. Predicted reactivity: Dog (84%), Pig (81%), Bovine (82%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Three different procarboxypeptidases A and two different procarboxypeptidases B have been isolated. The B1 and B2 forms differ from each other mainly in isoelectric point. Carboxypeptidase B1 is a highly tissue-specific protein and is a useful serum marker for acute pancreatitis and dysfunction of pancreatic transplants. It is not elevated in pancreatic carcinoma.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Carboxypeptidase B1
Polyclonal
Unconjugated
CPB1
0910001A18Rik; 1810063F02Rik; 2210008M23Rik; AI504870; carboxypeptidase B; carboxypeptidase B1; carboxypeptidase B1 (tissue); Cpb; Cpb1; Pancreas-specific protein; pancreatic carboxypeptidase B; pancreatic carboxypeptidase B1; PASP; PCPB; procarboxypeptidase B; protaminase; tissue carboxypeptidase B
Rabbit
Antigen affinity chromatography
RUO
1360, 24271, 76703
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
1 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
P15086, P19223
CPB1
Recombinant protein encompassing a sequence within the center region of human Carboxypeptidase B. The exact sequence is proprietary.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.