missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carboxypeptidase B1/CPB1 Antibody (008), DyLight 755, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Carboxypeptidase B1/CPB1 Monoclonal specifically detects Carboxypeptidase B1/CPB1 in Mouse samples. It is validated for Sandwich ELISA Detection.
Specifications
Specifications
| Antigen | Carboxypeptidase B1/CPB1 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 8 |
| Conjugate | DyLight 755 |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | carboxypeptidase B, carboxypeptidase B1 (tissue), CPB, DKFZp779K1333, EC 3.4.17, EC 3.4.17.2, Pancreas-specific protein, PASP, PCPB, procarboxypeptidase B |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Carboxypeptidase B1/CPB1 (NP_083982.1; Met1-Tyr415). |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?