missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Carboxypeptidase A4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568132
This item is not returnable.
View return policy
Description
Predicted to react with Mouse samples.
Carboxypeptidase A4 is a member of the carboxypeptidase A/B subfamily. Carboxypeptidases are zinc-containing exopeptidases that catalyze the release of carboxy-terminal amino acids, and are synthesized as zymogens that are activated by proteolytic cleavage. This protein could be involved in the histone hyperacetylation pathway. It is imprinted and may be a strong candidate protein for prostate cancer aggressiveness.
Specifications
| Carboxypeptidase A4 | |
| Polyclonal | |
| Unconjugated | |
| CPA4 | |
| 1110019K20Rik; AV009555; Carboxypeptidase A3; carboxypeptidase A4; CPA3; Cpa4; RGD1563619; UNQ694/PRO1339 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51200, 71791 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q6P8K8, Q9UI42 | |
| CPA4 | |
| Synthesized peptide derived from human Carboxypeptidase A4. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction