missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carboxypeptidase A4/CPA4 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£171.00 - £387.00
Specifications
| Antigen | Carboxypeptidase A4/CPA4 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18615190
|
Novus Biologicals
NBP2-92372-0.02ml |
0.02 mL |
£171.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18625190
|
Novus Biologicals
NBP2-92372-0.1ml |
0.1 mL |
£387.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Carboxypeptidase A4/CPA4 Polyclonal antibody specifically detects Carboxypeptidase A4/CPA4 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| Carboxypeptidase A4/CPA4 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology | |
| PBS with 50% glycerol, pH7.3. | |
| 51200 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Carboxypeptidase A3, carboxypeptidase A4, CPA3EC 3.4.17, EC 3.4.17.-, EC 3.4.17.1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 114-280 of human CPA4 (NP_057436.2). SSNNFNYGAYHSLEAIYHEMDNIAADFPDLARRVKIGHSFENRPMYVLKFSTGKGVRRPAVWLNAGIHSREWISQATAIWTARKIVSDYQRDPAITSILEKMDIFLLPVANPDGYVYTQTQNRLWRKTRSRNPGSSCIGADPNRNWNASFAGKGASDNPCSEVYHGP | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title