missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carboxypeptidase A3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16236-1-AP-20UL
This item is not returnable.
View return policy
Description
Three different forms of human pancreatic procarboxypeptidase A have been isolated. This gene encodes a form which is obtained as a binary complex of a procarboxypeptidase A with proproteinase E and functions as a secretory granule metalloexopeptidase.Specifications
| Carboxypeptidase A3 | |
| Polyclonal | |
| Unconjugated | |
| CPA3 | |
| Carboxypeptidase A3, CPA3, Mast cell carboxypeptidase A, MC CPA | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12873, 1359, 54242 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.26 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P15088, P15089, P21961 | |
| Cpa3 | |
| Carboxypeptidase A3 Fusion Protein Ag8984 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction