missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carboxypeptidase A1/CPA1 Antibody (CPA1/2712), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79880AF350
This item is not returnable.
View return policy
Description
Carboxypeptidase A1/CPA1 Monoclonal specifically detects Carboxypeptidase A1/CPA1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Carboxypeptidase A1/CPA1 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CPA1 | |
| Recombinant full-length human Carboxypeptidase A1/CPA1 protein (Uniprot: P15085) | |
| 0.1 mL | |
| Mast Cell Markers | |
| Human | |
| IgG2c κ |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CPA1/2712 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| carboxypeptidase A1, carboxypeptidase A1 (pancreatic), CPA, EC 3.4.17, EC 3.4.17.1 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur