missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carboxypeptidase A1/CPA1 Antibody (CL6607) [Alexa Fluor™ 700], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-76507AF700
This item is not returnable.
View return policy
Description
Carboxypeptidase A1/CPA1 Monoclonal antibody specifically detects Carboxypeptidase A1/CPA1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| Carboxypeptidase A1/CPA1 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| carboxypeptidase A1, carboxypeptidase A1 (pancreatic), CPA, EC 3.4.17, EC 3.4.17.1 | |
| This antibody was developed using a recombinant protein derived from P15085, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Mast Cell Markers | |
| 1357 | |
| Store at 4⪚C in the dark. | |
| IgG2b |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL6607 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction