missing translation for 'onlineSavingsMsg'
Learn More

Carboxymethylcellulose Sodium, TRC

Product Code. 30137689
Change view
Click to view available options
Quantity:
25 g
50 g
100 g
Unit Size:
100g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137689 100 g 100g
30138318 50 g 50g
17948234 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137689 Supplier Toronto Research Chemicals Supplier No. TRCC189548100G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9004-32-4
Molecular Formula C8H16NaO8
Molecular Weight (g/mol) 263.2
Synonym Accel 101, Accuwipe, Advantec D, Advice DP 27, Ahlstrom 939, Alicell LV, Alistaple 1168, Alpha Cel BH 100, Alphafloc, Alwall Plus 15, Amicon S 10Y1, Arbocel, Ariacel A 150, Armorib LS, Avicel, Avicel 101, Avicel 101 MCC, Avicel 102, Avicel 102330, Avicel 102331, Avicel 112, Avicel 2330, Avicel 2331, Avicel 591F, Avicel 955, Avicel AC 4125, Avicel CF 11, Avicel E 200, Avicel F 05, Avicel F 20, Avicel FD 100, Avicel FD 101, Avicel FD-F 20, Avicel GP 1030, Avicel GP 7210, Avicel HP 101, Avicel HP 105, Avicel LM 310, Avicel M 06, Avicel M 101, Avicel M 15, Avicel M 25, Avicel NT 020, Avicel NT 050, Avicel PA 200, Avicel PC 611, Avicel PH, Avicel PH 101, Avicel PH 101NF, Avicel PH 102, Avicel PH 102SCG, Avicel PH 103, Avicel PH 105, Avicel PH 112, Avicel PH 113, Avicel PH 200, Avicel PH 202, Avicel PH 301, Avicel PH 302, Avicel PH-F 10, Avicel PH-F 20, Avicel PH-M 06, Avicel PH-M 15, Avicel PH-M 25, Avicel R, Avicel RCS 91, Avicel SF, Avicel SP, Avicel TG 101, Avicel TG 101L, Avicel TG-D, Avicel TG-F 05, Avicel TG-F 20, BE 600-10TG, BGB Natural, BH 100, BH 40, BH 40 (Cellulose), BH 600/20 α-Cel, BMa 10002, BW 200, Bacell 569, Bahia SP, Bellfine D 10, BiNF-I-S, BiNFi-s AMA 100, BiNFi-s AMa 10002, BiNFi-s BMA 100, ‡BiNFi-s BMa 10002, BiNFi-s BMa 10010, BiNFi-s FMa 10002, BiNFi-s FMa 10005, BiNFi-s IMA 100, BiNFi-s IMA 1005, BiNFi-s IMa 10002, BiNFi-s MBa, BiNFi-s NFo 10002, BiNFi-s NMa 10002, BiNFi-s WFo 100, BiNFi-s WFo 10010, BiNFi-s WMa 100, BiNFi-s WMa 10002, BiNFi-s WMa 10005, BiNFi-s WMa 10010, BioPlus L, Borregaard 1200, Borregard Derivative HV, Borregard Derivative VHV, C 005F, C 010F, C 030, C 104843, C 104844, C 10583, C 30FM, C 500 (Cellulose), C 6663, C 8002, CA 107, CA 107-4W, CC 41, CF 1, CF 1 (Polysaccharide), CF 11, CFLL, CL 611, CNC, CNC-C, CNF 4, CNF-N 04, CP 102Y, CP 60, CPB 06, CR 1654, CRV, CRV (Polysaccharide), Celgum C 91;
IUPAC Name 2,3,4,5,6-pentahydroxyhexanal sodium salt (Polymer)
SMILES O=CC(O)C(O)C(O)C(O)CO.CC([O-])=O.[Na+]

Specifications

Quantity 100 g
InChI Formula InChI=1S/C6H12O6.C2H4O2.Na/c7-1-3(9)5(11)6(12)4(10)2-8;1-2(3)4;/h1,3-6,8-12H,2H2;1H3,(H,3,4);/q;;+1/p-1
Chemical Name or Material Carmellose Sodium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.