missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carboxylesterase 3/CES3/Esterase 31 Rabbit anti-Human, Clone: 372, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89803-50UL
This item is not returnable.
View return policy
Description
Carboxylesterase 3/CES3/Esterase 31 Monoclonal specifically detects Carboxylesterase 3/CES3/Esterase 31 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| Carboxylesterase 3/CES3/Esterase 31 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 23491 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA | |
| 372 | |
| Western Blot 1:500-1:2000, ELISA 1:5000-1:10000 | |
| carboxylesterase 3, carboxylesterase 3 (brain), EC 3.1.1, EC 3.1.1.1, ES31FLJ21736, esterase 31, Liver carboxylesterase 31 homolog | |
| Recombinant Human Carboxylesterase 3/CES3/Esterase 31 protein | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction