missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carboxylesterase 3/CES3/Esterase 31 Antibody (033), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-89802CL1
This item is not returnable.
View return policy
Description
Carboxylesterase 3/CES3/Esterase 31 Monoclonal antibody specifically detects Carboxylesterase 3/CES3/Esterase 31 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Carboxylesterase 3/CES3/Esterase 31 | |
| Monoclonal | |
| CoraFluor 1 | |
| carboxylesterase 3, carboxylesterase 3 (brain), EC 3.1.1, EC 3.1.1.1, ES31FLJ21736, esterase 31, Liver carboxylesterase 31 homolog | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Carboxylesterase 3/CES3/Esterase 31 (Accession#: NP_079198.2; Met1-Glu561). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| 033 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 23491 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction