missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carboxylesterase 1/CES1 Antibody (SR1990), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21940-50ul
This item is not returnable.
View return policy
Description
Carboxylesterase 1/CES1 Monoclonal antibody specifically detects Carboxylesterase 1/CES1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation
Specifications
| Carboxylesterase 1/CES1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Flow Cytometry, Immunoprecipitation | |
| SR1990 | |
| Western Blot 1:1000-1:5000, Flow Cytometry 1:100, Immunoprecipitation 1:40 | |
| ACAT, acyl coenzyme A:cholesterol acyltransferase, Acyl-coenzyme A:cholesterol acyltransferase, Brain carboxylesterase hBr1, carboxylesterase 1, carboxylesterase 1 (monocyte/macrophage serine esterase 1), carboxylesterase 2 (liver), CEH, Ces-1, CES1A2, CES2EC 3.1.1.1, cholesteryl ester hydrolase, Cocaine carboxylesterase, EC 3.1.1, egasyn, HMSE1, HMSECES1A1, human monocyte/macrophage serine esterase 1, liver carboxylesterase 1, MGC117365, Monocyte/macrophage serine esterase, PCE-1, REH, Retinyl ester hydrolase, Serine esterase 1, Ses-1, SES1TGH, Triacylglycerol hydrolase | |
| A synthesized peptide derived from human Carboxylesterase 1/CES1 (Uniprot #: P23141) | |
| 50 μg | |
| Cancer, Cardiovascular Biology, CD Markers, Endocrinology, Immunology, Signal Transduction | |
| 1066 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction