missing translation for 'onlineSavingsMsg'
Learn More

Carboxylesterase 1/CES1 Antibody [PerCP], Novus Biologicals™

Product Code. 30734338 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30734338 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30734338 Supplier Novus Biologicals Supplier No. AF7929C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Carboxylesterase 1/CES1 Polyclonal antibody specifically detects Carboxylesterase 1/CES1 in Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Carboxylesterase 1/CES1
Applications Western Blot
Classification Polyclonal
Conjugate PerCP
Formulation PBS
Gene Alias ACAT, acyl coenzyme A:cholesterol acyltransferase, Acyl-coenzyme A:cholesterol acyltransferase, Brain carboxylesterase hBr1, carboxylesterase 1, carboxylesterase 1 (monocyte/macrophage serine esterase 1), carboxylesterase 2 (liver), CEH, Ces-1, CES1A2, CES2EC 3.1.1.1, cholesteryl ester hydrolase, Cocaine carboxylesterase, EC 3.1.1, egasyn, HMSE1, HMSECES1A1, human monocyte/macrophage serine esterase 1, liver carboxylesterase 1, MGC117365, Monocyte/macrophage serine esterase, PCE-1, REH, Retinyl ester hydrolase, Serine esterase 1, Ses-1, SES1TGH, Triacylglycerol hydrolase
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Carboxylesterase 1/CES1, His19-Leu565, Accession # Q8VCC2
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cardiovascular Biology, CD Markers, Endocrinology, Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1066
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.