Learn More
Carboxy-PTIO potassium salt, 98+%
Beskrivning
Stable, water-soluble free radical that reacts with nitric oxide (NO) to inhibit NO synthase
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifikationer
Specifikationer
| Color | Blue |
| Quantity | 10 mg |
| Solubility Information | Soluble in water; Also soluble in ethanol,methanol,DMSO or DMF. |
| Formula Weight | 315.38 |
| Percent Purity | ≥98% |
| Physical Form | Solid |
| Chemical Name or Material | Carboxy-PTIO potassium salt |
RUO – Research Use Only
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.