missing translation for 'onlineSavingsMsg'
Learn More

Carboplatin, MedChemExpress

Product Code. p-200096621
Encompass_Preferred
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
This item is not returnable. View return policy

Product Code. 16491246

Brand: Medchem Express HY17393

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Carboplatin (NSC 241240) is a DNA synthesis inhibitor which binds to DNA, inhibits replication and transcription and induces cell death. Carboplatin (NSC 241240) is a derivative of CDDP and a potent anti-cancer agent.

Carboplatin (NSC 241240) is a DNA synthesis inhibitor which binds to DNA, inhibits replication and transcription and induces cell death. Carboplatin (NSC 241240) is a derivative of CDDP and a potent anti-cancer agent.

Specifications

Chemical Name or Material Carboplatin
Physical Form Powder
CAS 41575-94-4
Color White
Quantity 200 mg
Purity Grade Notes Research
Molecular Formula C6H12N2O4Pt
For Use With (Application) Cancer-programmed cell death
Synonym NSC 241240
Solubility Information H2O : 10.42 mg/mL (28.07 mM; ultrasonic and warming and heat to 60°C; DMSO can inactivate Carboplatin's activity) ∣DMF : 1 mg/mL (2.69 mM; Need ultrasonic; DMSO can inactivate Carboplatin's activity)
SMILES O=C1C2(CCC2)C(O[Pt]O1)=O.N.N
Molecular Weight (g/mol) 371.25
Formula Weight 371.25
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.