missing translation for 'onlineSavingsMsg'
Learn More

Carbonyldihydridotris(triphenylphosphine)ruthenium(II), 99%

Product Code. 11338518
Change view
Click to view available options
Quantity:
0.5 g
2 g
10 g
Unit Size:
0.5g
10g
2g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11338518 2 g 2g
11358518 0.5 g 0.5g
11348518 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11338518 Supplier Thermo Scientific Alfa Aesar Supplier No. 040524.04
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 25360-32-1 | C55H47OP3Ru | 917.97 g/mol

Carbonyldihydridotris(triphenylphosphine)ruthenium(II) is used as Catalyst for conversion of 1,4-alkynediols into pyrroles, Catalyst for oxidation of primary alcohols to methyl esters, Catalyst for rearrangement of oximes to amides, Catalyst for C-C coupling via directed C-H activation, Catalyst for enyne cyclization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25360-32-1
Molecular Formula C55H47OP3Ru
Molecular Weight (g/mol) 917.97
MDL Number MFCD00015870 MFCD00015870
InChI Key MLIYPCQSOXNTLJ-UHFFFAOYSA-N
PubChem CID 53384314
IUPAC Name carbon monoxide;hydride;ruthenium(2+);triphenylphosphane
SMILES [RuH2].[C-]#[O+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 161°C to 163°C
Color White
Assay Percent Range 99%
Quantity 2 g
Sensitivity Light and moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 917.97
Physical Form Crystals
Chemical Name or Material Carbonyldihydridotris(triphenylphosphine)ruthenium(II)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.