missing translation for 'onlineSavingsMsg'
Learn More

Carbonylchlorohydridotris(triphenylphosphine)ruthenium(II)

Product Code. 10782475 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10782475 1 g Glass bottle 1g
10782665 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10782475 Supplier Thermo Scientific Acros Supplier No. 392140010
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 16971-33-8 | C55H46ClOP3Ru | 952.41 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16971-33-8
Molecular Formula C55H46ClOP3Ru
Molecular Weight (g/mol) 952.41
MDL Number MFCD00049804
InChI Key IMPHYUNMDWWJAH-UHFFFAOYSA-M
Synonym carbonylchlorohydridotris triphenylphosphine-ruthenium ii
PubChem CID 71463878
IUPAC Name formaldehyde;hydride;ruthenium(2+);triphenylphosphane;chloride
SMILES Cl[RuH++].[C-]#[O+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 204.0°C to 206.0°C
Color Cream to Tan
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble
Formula Weight 952.4
Physical Form Powder
Chemical Name or Material Carbonylchlorohydridotris(triphenylphosphine)ruthenium(II)
compliance-icons
Product Identifier
  • Carbonylchlorohydridotris(triphenylphosphine)ruthenium(II)
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.