missing translation for 'onlineSavingsMsg'
Learn More

Carbonyl cyanide 3-chlorophenylhydrazone, 98%, Thermo Scientific Chemicals

Catalog Number 10165140
Click to view available options
Quantity:
100 mg
1 g
Packaging:
Glass bottle
555-60-2
C9H5ClN4
204.62
MFCD00001848
UGTJLJZQQFGTJD-UHFFFAOYSA-N
cccp, 3-chlorophenyl hydrazonomalononitrile, carbonylcyanide-3-chlorophenylhydrazone, carbonyl cyanide 3-chlorophenylhydrazone, carbonyl cyanide m-chlorophenyl hydrazone, carbonyl cyanide m-chlorophenylhydrazone, 3-chlorophenyl hydrazono malononitrile, m-chlorophenyl carbonylcyanide hydrazone, m-cl-ccp, carbonyl cyanide, 3-chlorophenyl hydrazone
2603
CHEBI:3259
2-[(3-chlorophenyl)hydrazinylidene]propanedinitrile
C1=CC(=CC(=C1)Cl)NN=C(C#N)C#N
555-60-2
C9H5ClN4
204.62
MFCD00001848
UGTJLJZQQFGTJD-UHFFFAOYSA-N
cccp, 3-chlorophenyl hydrazonomalononitrile, carbonylcyanide-3-chlorophenylhydrazone, carbonyl cyanide 3-chlorophenylhydrazone, carbonyl cyanide m-chlorophenyl hydrazone, carbonyl cyanide m-chlorophenylhydrazone, 3-chlorophenyl hydrazono malononitrile, m-chlorophenyl carbonylcyanide hydrazone, m-cl-ccp, carbonyl cyanide, 3-chlorophenyl hydrazone
2603
CHEBI:3259
2-[(3-chlorophenyl)hydrazinylidene]propanedinitrile
C1=CC(=CC(=C1)Cl)NN=C(C#N)C#N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 172°C to 177°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula ClC6H4NHN=C(CN)2
Quantity 1 g
Solubility Information Solubility in water: insoluble
Formula Weight 204.62
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Carbonyl cyanide 3-chlorophenylhydrazone, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • Carbonyl cyanide 3-chlorophenylhydrazone
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P311-Call a POISON CENTER/doctor/ .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'