missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Carbonic Anhydrase XIV Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA582355

Product Code. 16389314

  • £480.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: GQHWTYEGPH GQDHWPASYP ECGNNAQSPI DIQTDSVTFD PDLPALQPHG YDQPGTEPLD LHNNGHTVQL SLPSTLYLGG LPRKYVAAQL HLHWGQKGSP GGSEHQINSE ATFAELHIVH YDSDSYDSLS EAAERPQGLA VLGIL.

CA14 belongs a large family of zinc metalloenzymes that catalyze the versible hydration of carbon dioxide. The protein is predicted to be a type I membrane protein and shares highest sequence similarity with the other transmembrane CA isoform, CA XII; however, they have different patterns of tissue-specific expression and thus may play different physiologic roles.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Carbonic Anhydrase XIV
Polyclonal
Unconjugated
CA14
AW536446; CA XIV; CA14; Car14; Carbonate dehydratase XIV; carbonic anhydrase 14; Carbonic anhydrase XIV; carbonic dehydratase; Catm; CAXiV; CA-XIV; UNQ690/PRO1335
Rabbit
Antigen affinity chromatography
RUO
23632
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9ULX7
CA14
Recombinant protein corresponding to Human CA14. Recombinant protein control fragment (Product #RP-90144).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.