Learn More
Invitrogen™ Carbonic Anhydrase XIV Monoclonal Antibody (312908)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524035
Description
In direct ELISAs and Western blots, approximately 50% cross-reactivity with recombinant mouse CA14, 10% cross-reactivity with recombinant human (rh) CA5A, and no cross-reactivity with rhCA1, 2, 3, 4, 6, 7, 8, 9, 10, 12, or 13 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
CA14 belongs a large family of zinc metalloenzymes that catalyze the versible hydration of carbon dioxide. The protein is predicted to be a type I membrane protein and shares highest sequence similarity with the other transmembrane CA isoform, CA XII; however, they have different patterns of tissue-specific expression and thus may play different physiologic roles.
Specifications
| Carbonic Anhydrase XIV | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q9ULX7 | |
| CA14 | |
| Mouse myeloma cell line NS0-derived recombinant human Carbonic AnhydraseXIV/CA14 Gly19-Met290. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunoprecipitation, Western Blot | |
| 312908 | |
| Unconjugated | |
| CA14 | |
| AW536446; CA XIV; CA14; Car14; Carbonate dehydratase XIV; carbonic anhydrase 14; Carbonic anhydrase XIV; carbonic dehydratase; Catm; CAXiV; CA-XIV; UNQ690/PRO1335 | |
| Mouse | |
| Protein G | |
| RUO | |
| 23632 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.