missing translation for 'onlineSavingsMsg'
Learn More

Carbonic Anhydrase XIV/CA14 Antibody (515), DyLight 755, Novus Biologicals™

Product Code. 18337390 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18337390 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18337390 Supplier Novus Biologicals Supplier No. NBP289510IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Carbonic Anhydrase XIV/CA14 Monoclonal specifically detects Carbonic Anhydrase XIV/CA14 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Carbonic Anhydrase XIV/CA14
Applications ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 515
Conjugate DyLight 755
Formulation 50 mM Sodium Borate
Gene Alias Carbonate dehydratase XIV, carbonic anhydrase XIVcarbonic anhydrase 14, carbonic dehydratase, CAXiV, CA-XIV, EC 4.2.1.1
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Carbonic Anhydrase XIV/CA14 (NP_036245.1; Met 1-Met 290).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 23632
Test Specificity No cross-reactivity in ELISA with: Human CARPX Human CA12 Mouse CA14 Mouse, Rat
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.