missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbonic Anhydrase XIII Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539077
This item is not returnable.
View return policy
Description
Carbonic anhydrases (CAs) are a family of zinc metalloenzymes. For background information on the CA family, see MIM 114800.[supplied by OMIM, Mar 2008].
Specifications
| Carbonic Anhydrase XIII | |
| Polyclonal | |
| Unconjugated | |
| CA13 | |
| CAXIII, CA-XIII, carbonate dehydratase XIII, carbonic anhydrase 13 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 377677, 71934 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8N1Q1, Q9D6N1 | |
| CA13, Car13 | |
| A synthetic peptide derived from the internal region of human CA13 | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction