missing translation for 'onlineSavingsMsg'
Learn More

Carbonic Anhydrase XIII/CA13 Antibody, DyLight 680, Novus Biologicals™

Product Code. 18365482 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18365482 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18365482 Supplier Novus Biologicals Supplier No. NBP299317FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Carbonic Anhydrase XIII/CA13 Polyclonal specifically detects Carbonic Anhydrase XIII/CA13 in Human samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Carbonic Anhydrase XIII/CA13
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate DyLight 680
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias carbonic anhydrase 13, carbonic anhydrase XIIICarbonate dehydratase XIII, CAXIII, CA-XIII, EC 4.2.1.1, FLJ37995, MGC59868
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Carbonic Anhydrase XIII/CA13 (NP_940986.1; Met 1-His 262).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 377677
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.