missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbonic Anhydrase XIII/CA13 Antibody (103), mFluor Violet 450 SE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Carbonic Anhydrase XIII/CA13 Monoclonal antibody specifically detects Carbonic Anhydrase XIII/CA13 in Human samples. It is validated for ELISA
Specifications
Specifications
| Antigen | Carbonic Anhydrase XIII/CA13 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 103 |
| Conjugate | mFluor Violet 450 SE |
| Dilution | ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | carbonic anhydrase 13, carbonic anhydrase XIIICarbonate dehydratase XIII, CAXIII, CA-XIII, EC 4.2.1.1, FLJ37995, MGC59868 |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human Carbonic Anhydrase XIII/CA13 (Accession#: NP_940986.1; Met1-His262). |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?