missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Carbonic Anhydrase XII Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA597528
This item is not returnable.
View return policy
Description
Carbonic Anhydrase XII Polyclonal Antibody for Western Blot, IHC (P), ELISA
Carbonic Anhydrase XII is a protein coding gene.
Specifications
| Carbonic Anhydrase XII | |
| Polyclonal | |
| Unconjugated | |
| CA12 | |
| 2310047E01Rik; AI314958; CA12; Car12; Carbonate dehydratase XII; carbonic anhydrase 12; Carbonic anhydrase XII; carbonic anyhydrase 12; carbonic dehydratase; CAXII; CA-XII; HsT18816; membrane-bound carbonic anhydrase 12; T18816; tumor antigen HOM-RCC-3.1.3 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 76459, 771 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 3 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| O43570, Q8CI85 | |
| CA12, Car12 | |
| Recombinant Human Carbonic anhydrase 12 protein (25-301AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction