missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Carbonic Anhydrase XII Polyclonal Antibody
GREENER_CHOICE

Product Code. 15577792
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15577792 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15577792 Supplier Invitrogen™ Supplier No. PA547303

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

In direct ELISAs and Western blots, approximately 40% cross-reactivity with recombinant human CA12 is observed and less than 5% cross-reactivity with recombinant human CA1, rhCA2, rhCA3, rhCA8, recombinant mouse (rm) CA4, and rmCA9 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

Carbonic Anhydrase XII is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Carbonic Anhydrase XII
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Car12
Gene Accession No. Q8CI85
Gene Alias 2310047E01Rik; AI314958; CA12; Car12; Carbonate dehydratase XII; carbonic anhydrase 12; Carbonic anhydrase XII; carbonic anyhydrase 12; carbonic dehydratase; CAXII; CA-XII; HsT18816; membrane-bound carbonic anhydrase 12; T18816; tumor antigen HOM-RCC-3.1.3
Gene Symbols Car12
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Carbonic Anhydrase XII/CA12 Ala25-Ser301.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 76459
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.