missing translation for 'onlineSavingsMsg'
Learn More

Carbonic Anhydrase XII/CA12 Antibody (001), DyLight 650 , Novus Biologicals™

Product Code. 18422239 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18422239 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18422239 Supplier Novus Biologicals Supplier No. NBP290350C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Carbonic Anhydrase XII/CA12 Monoclonal antibody specifically detects Carbonic Anhydrase XII/CA12 in Mouse samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Carbonic Anhydrase XII/CA12
Applications ELISA
Classification Monoclonal
Clone 001
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias Carbonate dehydratase XII, carbonic anhydrase 12, carbonic anhydrase XIICAXII, carbonic dehydratase, CA-XII, EC 4.2.1.1, FLJ20151, HsT18816, Tumor antigen HOM-RCC-3.1.3
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Carbonic Anhydrase XII/CA12 (NP_848483.2; Met1-Ser301).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 771
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.