missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Carbonic Anhydrase X Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563029
This item is not returnable.
View return policy
Description
Immunogen sequence: HRLEEIRLHF GSEDSQGSEH LLNGQAFSGE VQLIHYNHEL YTNVTEAAKS PNGLVVVSIF IKVSDSSNPF LNRMLNR Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
This gene encodes a protein that belongs to the carbonic anhydrase family of zinc metalloenzymes, which catalyze the reversible hydration of carbon dioxide in various biological processes. The protein encoded by this gene is an acatalytic member of the alpha-carbonic anhydrase subgroup, and it is thought to play a role in the central nervous system, especially in brain development. Multiple transcript variants encoding the same protein have been found for this gene.
Specifications
| Carbonic Anhydrase X | |
| Polyclonal | |
| Unconjugated | |
| CA10 | |
| 2700029L05Rik; BB085816; Ca10; Car10; carbonic anhydrase 10; carbonic anhydrase X; carbonic anhydrase-related protein 10; Carbonic anhydrase-related protein X; CARP X; CA-RP X; CARPX; CA-RPX; Cerebral protein 15; cerebral protein-15; HUCEP-15; UNQ533/PRO1076 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 56934 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NS85 | |
| CA10 | |
| Recombinant protein corresponding to Human Carbonic Anhydrase X. Recombinant protein control fragment (Product #RP-101386). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction