missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbonic Anhydrase VIII Polyclonal Antibody
Carbonic Anhydrase VIII Polyclonal Antibody
Brand: Invitrogen PA547267
This item is not returnable.
View return policy
Description
Carbonic Anhydrase VIII Polyclonal Antibody for Western Blot, IP
Specifications
| Carbonic Anhydrase VIII | |
| Polyclonal | |
| Unconjugated | |
| Ca8 | |
| AW546993; Ca8; Cals; Cals1; CAMRQ3; Car8; carbonate dehydratase; carbonic anhydrase 8; carbonic anhydrase VIII; carbonic anhydrase-like sequence; carbonic anhydrase-like sequence 1; carbonic anhydrase-related protein; CA-related protein; CARP; CA-RP; CARP VIII; CA-RP VIII; CA-VIII; hypothetical protein LOC550233; LOW QUALITY PROTEIN: carbonic anhydrase-related protein; MGC120502; MGC99509; wdl; zgc:110118 | |
| Goat | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12319, 767 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunoprecipitation, Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| P28651, P35219 | |
| Ca8, Car8 | |
| E. coli-derived recombinant human Carbonic AnhydraseVIII/CA8 Ala2-Gln290. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction