missing translation for 'onlineSavingsMsg'
Learn More

Carbonic Anhydrase VIII/CA8 Antibody (CA8/6572), DyLight 680, Novus Biologicals™

Product Code. 18383202
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18383202 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18383202 Supplier Bio-Techne Supplier No. NBP314030FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Carbonic Anhydrase VIII/CA8 Monoclonal antibody specifically detects Carbonic Anhydrase VIII/CA8 in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Carbonic Anhydrase VIII/CA8
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CA8/6572
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias CALScarbonate dehydratase, CAMRQ3, carbonic anhydrase VIIIMGC99509, carbonic anhydrase-like sequence, carbonic anhydrase-related protein, CARPCA-related protein, CA-VIII, MGC120502
Host Species Mouse
Immunogen Recombinant full-length human Carbonic Anhydrase VIII/CA8 protein (Uniprot: P35219)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 767
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.