missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbonic Anhydrase VIII/CA8 Antibody (001), Alexa Fluor™ 700, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Carbonic Anhydrase VIII/CA8 Monoclonal specifically detects Carbonic Anhydrase VIII/CA8 in Mouse samples. It is validated for ELISA, Sandwich ELISA Detection.
Specifications
Specifications
| Antigen | Carbonic Anhydrase VIII/CA8 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 1 |
| Conjugate | Alexa Fluor 700 |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | CALScarbonate dehydratase, CAMRQ3, carbonic anhydrase VIIIMGC99509, carbonic anhydrase-like sequence, carbonic anhydrase-related protein, CARPCA-related protein, CA-VIII, MGC120502 |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Carbonic Anhydrase VIII/CA8 (P28651; Met1-Gln291). |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?