missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Carbonic Anhydrase VII/CA7 Monoclonal antibody specifically detects Carbonic Anhydrase VII/CA7 in Human samples. It is validated for Western Blot, ELISA
Specifications
Specifications
| Antigen | Carbonic Anhydrase VII/CA7 |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | 3B7 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500 |
| Formulation | In 1x PBS, pH 7.4 |
| Gene Accession No. | NP_005173 |
| Gene Alias | Carbonate dehydratase VII, carbonic anhydrase 7, carbonic anhydrase VIICAVII, carbonic dehydratase VII, CA-VII, EC 4.2.1.1 |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?