missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Carbonic Anhydrase VI Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA583027

Product Code. 16369984

  • £443.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: SEISGSEHTV DGIRHVIEIH IVHYNSKYKS YDIAQDAPDG LAVLAAFVEV KNYPENTYYS NFISHLANIK YPGQRTTLTG LDVQDMLPRN LQHYYTY.

The protein encoded by this gene is one of several isozymes of carbonic anhydrase. This protein is found only in salivary glands and saliva and protein may play a role in the reversible hydratation of carbon dioxide though its function in saliva is unknown.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Carbonic Anhydrase VI
Polyclonal
Unconjugated
CA6
CA6; CA-6; Car6; Carbonate dehydratase VI; carbonic anhydrase 6; Carbonic anhydrase VI; carbonic anhydrase VI nirs variant 2; Carbonic anhydrase6; CA-VI; DOC1; GUSTIN; salivary carbonic anhydrase; Secreted carbonic anhydrase
Rabbit
Antigen Affinity Chromatography
RUO
765
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
P23280
CA6
Recombinant protein corresponding to Human CA6. Recombinant protein control fragment (Product #RP-94191).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.