missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Carbonic Anhydrase VI Monoclonal Antibody (401819)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524137
This item is not returnable.
View return policy
Description
In Western blots, no cross-reactivity with recombinant human Carbonic Anhydrase 2, 3, 4, 5A, 5B, 7, 8, 9, recombinant mouse Carbonic Anhydrase 12, and 14 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
The protein encoded by this gene is one of several isozymes of carbonic anhydrase. This protein is found only in salivary glands and saliva and protein may play a role in the reversible hydratation of carbon dioxide though its function in saliva is unknown.
Specifications
| Carbonic Anhydrase VI | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| P23280 | |
| CA6 | |
| Mouse myeloma cell line NS0-derived recombinant human Carbonic Anhydrase VI Gln18-Asn308. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunoprecipitation, Western Blot | |
| 401819 | |
| Unconjugated | |
| CA6 | |
| CA6; CA-6; Car6; Carbonate dehydratase VI; carbonic anhydrase 6; Carbonic anhydrase VI; carbonic anhydrase VI nirs variant 2; Carbonic anhydrase6; CA-VI; DOC1; GUSTIN; salivary carbonic anhydrase; Secreted carbonic anhydrase | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 765 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction