missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbonic Anhydrase VI Monoclonal Antibody (401809)
Carbonic Anhydrase VI Monoclonal Antibody (401809)
Brand: Invitrogen MA524136
This item is not returnable.
View return policy
Description
Specifications
| Carbonic Anhydrase VI | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| P23280 | |
| CA6 | |
| Mouse myeloma cell line NS0-derived recombinant human Carbonic Anhydrase VI Gln18-Asn308. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Flow Cytometry | |
| 401809 | |
| Unconjugated | |
| CA6 | |
| CA6; CA-6; Car6; Carbonate dehydratase VI; carbonic anhydrase 6; Carbonic anhydrase VI; carbonic anhydrase VI nirs variant 2; Carbonic anhydrase6; CA-VI; DOC1; GUSTIN; salivary carbonic anhydrase; Secreted carbonic anhydrase | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 765 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction