missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Carbonic Anhydrase VB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA525944
This item is not returnable.
View return policy
Description
This antibody is predicted to react with rat based on sequence homology.
Carbonic anhydrases (CAs) are members of a large family of zinc metalloenzymes responsible for catalyzing the reversible hydration of carbon dioxide.CAs show extensive diversity in their distribution and subcellular localization.They are involved in a variety of biological processes, including calcification, bone resorption, respiration, acid-base balance and the formation of aqueous humor, saliva, gastric juice and cerebrospinal fluid. CA VB, also known as carbonate dehydratase VB, is one of two isoforms of CA V. It localizes to the mitochondria and is involved in metabolic processes. CA VB is predominantly expressed in heart, pancreas, lung, placenta, kidney and skeletal muscle. It exhibits highest homology with family member CA VA (the second isoform of CA V); however, unlike CA VA, it is not expressed in the liver, suggesting that it plays a significantly different physiological role.
Specifications
| Carbonic Anhydrase VB | |
| Polyclonal | |
| Unconjugated | |
| CA5B | |
| 7330410H16Rik; Ca5b; Car5b; Carbonate dehydratase VB; carbonic anhydrase 5B; carbonic anhydrase 5b, mitochondrial; Carbonic anhydrase VB; carbonic anhydrase VB, mitochondrial; carbonic dehydratase; CarVb; CAVB; CA-VB; D730005F19Rik | |
| Rabbit | |
| Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
| RUO | |
| 11238, 56078 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 2 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q9QZA0, Q9Y2D0 | |
| CA5B, Car5b | |
| KLH conjugated synthetic peptide between 253-283 amino acids from the C-terminal region of human CA5B. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction