missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Carbonic Anhydrase VB Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA552959

Product Code. 15720911

  • £467.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: GLAVIGVFLK LGKHHKELQK LVDTLPSIKH KDALVEFGSF DPSCLMPTCP DYWTYSGSLT TPPLSESVTW IIKKQPVEVD HDQLEQFRTL LFTSEGEKEK RMVD Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.

Carbonic anhydrases (CAs) are members of a large family of zinc metalloenzymes responsible for catalyzing the reversible hydration of carbon dioxide.CAs show extensive diversity in their distribution and subcellular localization.They are involved in a variety of biological processes, including calcification, bone resorption, respiration, acid-base balance and the formation of aqueous humor, saliva, gastric juice and cerebrospinal fluid. CA VB, also known as carbonate dehydratase VB, is one of two isoforms of CA V. It localizes to the mitochondria and is involved in metabolic processes. CA VB is predominantly expressed in heart, pancreas, lung, placenta, kidney and skeletal muscle. It exhibits highest homology with family member CA VA (the second isoform of CA V); however, unlike CA VA, it is not expressed in the liver, suggesting that it plays a significantly different physiological role.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Carbonic Anhydrase VB
Polyclonal
Unconjugated
CA5B
7330410H16Rik; Ca5b; Car5b; Carbonate dehydratase VB; carbonic anhydrase 5B; carbonic anhydrase 5b, mitochondrial; Carbonic anhydrase VB; carbonic anhydrase VB, mitochondrial; carbonic dehydratase; CarVb; CAVB; CA-VB; D730005F19Rik
Rabbit
Antigen Affinity Chromatography
RUO
11238
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9Y2D0
CA5B
Recombinant protein corresponding to Human Carbonic Anhydrase VB. Recombinant protein control fragment (Product #RP-92233).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.