missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Carbonic Anhydrase VA Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA589996

Product Code. 16577641

  • £441.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: CAWQTSNNTL HPLWTVPVSV PGGTRQSPIN IQWRDSVYDP QLKPLRVSYE AASCLYIWNT GYLFQVEFDD ATEASGISGG PLENHYRLKQ FHFHWGAVNE GGSEHTVDGH AYPAELHLVH WNSVKYQNYK EAVVGENGLA VIGVFLKLGA HHQTLQRLVD ILPEIKHKDA RAAMRPFDPS TLLPTCWDYW TYAGSLTTPP LTESVTWIIQ KEPVEVAPSQ LSAFRTLLFS ALGEEEKMMV NNYRPLQPLM NRKVWASFQA TNEGTRS; Positive Samples: K-562, LO2, HepG2; Cellular Location: Mitochondrion.

Carbonic anhydrases (CAs) are members of a large family of zinc metalloenzymes that catalyze the reversible hydration of carbon dioxide. CAs are involved in a variety of biological processes, including respiration, calcification, acid-base balance, bone resorption, and the formation of aqueous humor, cerebrospinal fluid, saliva, and gastric juice. They show extensive diversity in distribution and in their subcellular localization. The human CA5A gene encodes for the CA V protein which is expressed in the mitochondrial matrix in liver tissues, but can also be detected in many other tissues. In ureagenesis, the CA V protein may be involved in providing bicarbonate to carbamoyl phosphate synthetase.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Carbonic Anhydrase VA
Polyclonal
Unconjugated
Car5a
CA Y; CA5; CA5A; CA5AD; Car5; Car5a; Carbonate dehydratase VA; carbonic anhydrase 5 mitochondrial; carbonic anhydrase 5, mitochondrial; carbonic anhydrase 5A; carbonic anhydrase 5a, mitochondrial; carbonic anhydrase 5b, mitochondrial; carbonic anhydrase V, mitochondrial; carbonic anhydrase VA; carbonic anhydrase VA, mitochondrial; carbonic dehydratase; CAV; CAVA; CA-VA; GS1-21A4.1
Rabbit
Affinity chromatography
RUO
12352
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot, Immunocytochemistry
2.05 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
P23589
Car5a
Recombinant fusion protein containing a sequence corresponding to amino acids 39-305 of human CA5A (NP_0017301).
100 μL
Primary
Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.