missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbonic Anhydrase VA/CA5A Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92181-0.02ml
This item is not returnable.
View return policy
Description
Carbonic Anhydrase VA/CA5A Polyclonal antibody specifically detects Carbonic Anhydrase VA/CA5A in Mouse samples. It is validated for Western BlotSpecifications
| Carbonic Anhydrase VA/CA5A | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| CA5carbonic anhydrase 5A, mitochondrial, Carbonate dehydratase VA, carbonic anhydrase V, mitochondrial, Carbonic anhydrase VA, carbonic anhydrase VA, mitochondrial, carbonic dehydratase, CAV, CAVA, CA-VA, EC 4.2.1.1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 39-305 of human CA5A (NP_001730.1). CAWQTSNNTLHPLWTVPVSVPGGTRQSPINIQWRDSVYDPQLKPLRVSYEAASCLYIWNTGYLFQVEFDDATEASGISGGPLENHYRLKQFHFHWGAVNEGGSEHTVDGHAYPAELHLVHWNSVKYQNYKEAVVGENGLAVIGVFLKLGAHHQTLQRLVDILPEIKHKDARAAMRPFDPSTLLPTCWDYWTYAGSLTTPPLTESVTWIIQKEPVEVAPSQLSAFRTLLFSALGEEEKMMVNNYRPLQPLMNRKVWASFQATNEGTRS | |
| 0.02 mL | |
| Lipid and Metabolism | |
| 763 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction