missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbonic Anhydrase IX/CA9 Mouse, Clone: SPM314, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP2-34360-0.02MG
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Carbonic Anhydrase IX/CA9 Monoclonal specifically detects Carbonic Anhydrase IX/CA9 in Human, Equine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Especificaciones
| Carbonic Anhydrase IX/CA9 | |
| Monoclonal | |
| Unconjugated | |
| PBS with 0.05% BSA. with 0.05% Sodium Azide | |
| CA-IX, CAIXcarbonic anhydrase 9, Carbonate dehydratase IX, carbonic anhydrase IXpMW1, carbonic dehydratase, EC 4.2.1.1, G250, Membrane antigen MN, MNRCC-associated antigen G250, P54/58N, RCC-associated protein G250, Renal cell carcinoma-associated antigen G250 | |
| Mouse | |
| 200 kDa | |
| 0.02 mg | |
| Cancer, Cellular Markers, HIF Target Genes, Hypoxia, Lipid and Metabolism, Signal Transduction, Vision | |
| 768 | |
| Human, Equine | |
| Purified |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| SPM314 | |
| Western Blot 0.5-1ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 1-2ug/ml, Immunoprecipitation 1-2ug/500ug, Immunohistochemistry-Paraffin 0.5-1ug/ml, Immunohistochemistry-Frozen 0.5-1.0ug/ml | |
| Q16790 | |
| CA9 | |
| Microsomal fraction of human renal cortical tissue homogenate | |
| Protein A purified | |
| RUO | |
| Primary | |
| Recognizes a glycoprotein of ∼200kDa, identified as carbonic anhydrase IX (CAIX/gp200). Its epitope resides in the carbohydrate domain of gp200. It shows no significant cross-reactivity with other carbohydrate determinants, such as the Lewis blood group antigens, epithelial membrane antigen, HMFG, and AB blood group antigens. In normal kidney, gp200 is localized along the brush border of the pars convoluta and pars recta segments of the proximal tubule, as well as focally along the luminal surface of Bowman's capsule adjoining the outgoing proximal tubule. Reportedly, gp200 is expressed by 93% of primary and 84% of metastatic renal cell carcinomas. This MAb may be useful in the investigations of carcinomas of proximal nephrogenic differentiation especially those showing tubular differentiation. | |
| Store at 4C. | |
| IgG2b κ |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido