missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbonic Anhydrase IX/CA9 Antibody (CA9/781), Novus Biologicals™
Mouse Monoclonal Antibody
£211.00 - £373.00
Specifications
| Antigen | Carbonic Anhydrase IX/CA9 |
|---|---|
| Clone | CA9/781 |
| Concentration | 0.2mg/mL |
| Dilution | Western Blot 0.5 - 1.0 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, SDS-Page, Immunofluorescence 1 - 2 ug/ml |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), SDS-Page, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18118227
|
Novus Biologicals
NBP2-45237-0.02MG |
0.02 mg |
£211.00
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18185168
|
Novus Biologicals
NBP2-45237-0.1MG |
0.1 mg |
£373.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ensure accurate, reproducible results in Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence
Carbonic Anhydrase IX/CA9 Monoclonal specifically detects Carbonic Anhydrase IX/CA9 in Human, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Carbonic Anhydrase IX/CA9 | |
| 0.2mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), SDS-Page, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Cancer, Cellular Markers, HIF Target Genes, Hypoxia, Lipid and Metabolism, Signal Transduction, Vision | |
| 10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
| CA-IX, CAIXcarbonic anhydrase 9, Carbonate dehydratase IX, carbonic anhydrase IXpMW1, carbonic dehydratase, EC 4.2.1.1, G250, Membrane antigen MN, MNRCC-associated antigen G250, P54/58N, RCC-associated protein G250, Renal cell carcinoma-associated antigen G250 | |
| CA9 | |
| IgG2b κ | |
| Protein A or G purified | |
| Recognizes a glycoprotein of ∼200kDa, identified as carbonic anhydrase IX (CAIX/gp200). Carbonic Anhydrases (CAs) are members of a large family of zinc metallo-enzymes that catalyze the reversible hydration of carbon dioxide. CAs are involved in a variety of biological processes, including respiration, calcification, acid-base balance, bone resorption and the formation of aqueous humor, cerebrospinal fluid, saliva and gastric juice. They show extensive diversity in distribution and in their subcellular localization. CA IX is specifically expressed in clear-cell renal carcinomas. |
| CA9/781 | |
| Western Blot 0.5 - 1.0 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, SDS-Page, Immunofluorescence 1 - 2 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Equine | |
| Q16790 | |
| 768 | |
| Recombinant human CA9 protein | |
| Primary | |
| Store at 4C. | |
| 55 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title