missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD32b Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA586930
This item is not returnable.
View return policy
Description
Purity is > 95% by SDS-PAGE.
This gene encodes one member of a family of immunoglobulin Fc receptor genes found on the surface of many immune response cells. The protein encoded by this gene is a cell surface receptor found on phagocytic cells such as macrophages and neutrophils, and is involved in the process of phagocytosis and clearing of immune complexes. Alternative splicing results in multiple transcript variants.
Specifications
| CD32b | |
| Polyclonal | |
| Unconjugated | |
| FCGR2B | |
| AI528646; CD32; CD32 receptor 2; CD32 receptor 2a; CD32 receptor 2b; CD32A; CD32B; CDw32; F630109E10Rik; Fc fragment of IgG receptor IIb; Fc fragment of IgG, low affinity II, receptor for (CD32); Fc fragment of IgG, low affinity IIb, receptor; Fc fragment of IgG, low affinity IIb, receptor (CD32); Fc fragment of IgG, low affinity IIb, receptor for (CD32); fc gamma receptor IIB; Fc gamma RIIb; Fc receptor, IgG, low affinity IIb; Fc[g]RII; FCG2; FC-gamma RII; fc-gamma RII-a; Fc-gamma RII-b; Fc-gamma-RII; fc-gamma-RIIa; FcgammaRIIB; fc-gamma-RIIb; FcgammaRIIB2; FcGR; Fcgr2; Fcgr2a; FCGR2A1; FCGR2B; FcgRII; Fcr-2; Fcr-3; fcRII; fcRII-a; fcRII-b; FGFR2B; IGFR2; IgG Fc receptor II beta; igG Fc receptor II-b; Low affinity immunoglobulin gamma Fc region receptor II; low affinity immunoglobulin gamma Fc region receptor II-b; Ly-17; LyM-1; Ly-m20; Lymphocyte antigen 17 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 14130, 2213, 289211 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P08101, P31994, Q63203 | |
| FCGR2B | |
| Synthetic peptide, corresponding to amino acids 250-300 of Human FCGR2B. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction