missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Carbonic Anhydrase IV Monoclonal Antibody (C16)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA541559
This item is not returnable.
View return policy
Description
Carbonic Anhydrase IV Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
Carbonic anhydrases are a large family of zinc metalloenzymes that catalyze the reversible hydration of carbon dioxide. They participate in a variety of biological processes, including respiration, calcification, acid-base balance, bone resorption, and the formation of aqueous humor, cerebrospinal fluid, saliva, and gastric acid. They show extensive diversity in tissue distribution and in their subcellular localization. This gene encodes a glycosylphosphatidyl-inositol-anchored membrane isozyme expressed on the luminal surfaces of pulmonary capillaries and proximal renal tubules. Its exact function is not known, however, it may have a role in inherited renal abnormalities of bicarbonate transport.
Specifications
| Carbonic Anhydrase IV | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P22748, P48284 | |
| CA4 | |
| Recombinant protein Carbonic Anhydrase IV. The antigen corresponds to amino acid range 19-283 of the target protein. | |
| 200 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG2b κ |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| C16 | |
| Unconjugated | |
| CA4 | |
| AW456718; CA IV; CA4; CAIV; CA-IV; Car4; Carbonate dehydratase IV; carbonic anhydrase 4; Carbonic anhydrase IV; carbonic dehydratase IV; RP17 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 29242, 762 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction