missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Carbonic Anhydrase IV Monoclonal Antibody (C16)

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA543925

 View more versions of this product

Product Code. 17694376

  • £177.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Carbonic Anhydrase IV Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

Carbonic anhydrases are a large family of zinc metalloenzymes that catalyze the reversible hydration of carbon dioxide. They participate in a variety of biological processes, including respiration, calcification, acid-base balance, bone resorption, and the formation of aqueous humor, cerebrospinal fluid, saliva, and gastric acid. They show extensive diversity in tissue distribution and in their subcellular localization. This gene encodes a glycosylphosphatidyl-inositol-anchored membrane isozyme expressed on the luminal surfaces of pulmonary capillaries and proximal renal tubules. Its exact function is not known, however, it may have a role in inherited renal abnormalities of bicarbonate transport.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Carbonic Anhydrase IV
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
P22748, P48284
CA4
Recombinant protein Carbonic Anhydrase IV. The antigen corresponds to amino acid range 19-283 of the target protein.
20 μL
Primary
Human, Rat
Antibody
IgG2b κ
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
C16
Unconjugated
CA4
AW456718; CA IV; CA4; CAIV; CA-IV; Car4; Carbonate dehydratase IV; carbonic anhydrase 4; Carbonic anhydrase IV; carbonic dehydratase IV; RP17
Mouse
Protein A/G
RUO
29242, 762
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.