missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbonic Anhydrase IV/CA4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-69435
This item is not returnable.
View return policy
Description
Carbonic Anhydrase IV/CA4 Polyclonal specifically detects Carbonic Anhydrase IV/CA4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Carbonic Anhydrase IV/CA4 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| CAIV, CA-IV, Car4, Carbonate dehydratase IV, carbonic anhydrase 4, carbonic anhydrase IVRP17, carbonic dehydratase IV, EC 4.2.1.1, retinitis pigmentosa 17 (autosomal dominant) | |
| Rabbit | |
| 34 kDa | |
| 100 μL | |
| Lipid and Metabolism, Vision | |
| 762 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| P22748 | |
| CA4 | |
| Synthetic peptides corresponding to CA4(carbonic anhydrase IV) The peptide sequence was selected from the C terminal of Carbonic Anhydrase IV. Peptide sequence AFSQKLYYDKEQTVSMKDNVRPLQQLGQRTVIKSGAPGRPLPWALPALLG. | |
| Affinity purified | |
| RUO | |
| Primary | |
| Rat: 85%; Mouse: 85%; Equine: 83%; Bovine: 83%. | |
| Human, Mouse, Rat, Bovine, Equine, Guinea Pig, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction