missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbonic Anhydrase IV/CA4 Antibody (039), Alexa Fluor™ 532, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89518AF532
This item is not returnable.
View return policy
Description
Carbonic Anhydrase IV/CA4 Monoclonal specifically detects Carbonic Anhydrase IV/CA4 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Carbonic Anhydrase IV/CA4 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| CAIV, CA-IV, Car4, Carbonate dehydratase IV, carbonic anhydrase 4, carbonic anhydrase IVRP17, carbonic dehydratase IV, EC 4.2.1.1, retinitis pigmentosa 17 (autosomal dominant) | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Carbonic Anhydrase IV/CA4 (NP_000708.1; Met1-Lys283). | |
| 0.1 mL | |
| Lipid and Metabolism, Vision | |
| 762 | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 39 | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| No cross-reactivity in ELISA with: Human CA2 Human CA3 Human CA5A Human CA5B Human CA8 Human CA9 Human CA10 Human CA12 Human CA13 Human CA14 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction